Learn More
Echelon Biosciences Research Labs 4-Diphosphocytidyl-2-C-methyl-D-erythritol (CDP-ME) 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs IM0521MG
Encompass
4-Diphosphocytidyl-2-C-methyl-D-erythritol (CDP-ME) is an intermediate in the non-mevalonate pathway. It is formed from MEP by CDPME Synthase and phosphorylated at the 2-position by CDPME kinase to CDP-MEP. This item is non returnable
Catalog No. 50-720-403