Learn More
Sigma Aldrich Fine Chemicals Biosciences CDP StarR Disodium 2 chlor
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11685627001
Encompass
Contents25 mM SOLUTIONon (12.38 mg/ml) 100x concentration
Catalog No. 50-290-3092