Learn More
TARGETMOL CHEMICALS INC CDPPB 10MG
SDP
Catalog No. 502580938
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDPPB is a modulator of mGluR5 positive allosteric(with an EC50 of 27 nM in Chinese hamster ovary cells expressing human mGluR5). purity: 99%
Catalog No. 50-258-0938 Supplier TARGETMOL CHEMICALS INC Supplier No. T2264110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDPPB is a modulator of mGluR5 positive allosteric(with an EC50 of 27 nM in Chinese hamster ovary cells expressing human mGluR5). purity: 99%