Learn More
Abcam CDPPB, positive allosteric modulator for mGlu5, 10MG
Catalog No. 502707654
Encompass_Preferred
MW 364.4 Da, Purity >98%. Potent, selective, positive allosteric modulator for mGlu5. In CHO cells expressing mGlu5 receptors, potentiates threshold responses to glutamate in fluorometric Ca2+ assays more than 7-fold (EC50 approximately 27 nM). Acts as mGlu5 agonist at higher concentrations. Brain penetrant and antipsychotic *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7654 Supplier Abcam Supplier No. AB12036310MG
MW 364.4 Da, Purity >98%. Potent, selective, positive allosteric modulator for mGlu5. In CHO cells expressing mGlu5 receptors, potentiates threshold responses to glutamate in fluorometric Ca2+ assays more than 7-fold (EC50 approximately 27 nM). Acts as mGlu5 agonist at higher concentrations. Brain penetrant and antipsychotic *in vivo*.
The product is subject to the following: Abcam Restricted Use Statement