Learn More
Aviva Systems Biology Corp CDR2L Antibody - N-terminal region (ARP39113_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39113_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDR2L. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CDR2L. Alternate names: HUMPPAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-783