Learn More
Aviva Systems Biology Corp CDRT4 Antibody - middle region (ARP55668_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55668_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDRT4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CDRT4. Alternate names: FLJ36674, MGC33988, NBLA10383ApplicationWB. Species reactivity: Human
Catalog No. 50-014-61572