Learn More
ABclonal Technology CDSN Rabbit pAb
SDP
Supplier:  ABclonal Technology A146021000UL
Encompass_Preferred
This gene encodes a protein found in corneodesmosomes, which localize to human epidermis and other cornified squamous epithelia. The encoded protein undergoes a series of cleavages during corneocyte maturation. This gene is highly polymorphic in human populations, and variation has been associated with skin diseases such as psoriasis, hypotrichosis and peeling skin syndrome. The gene is located in the major histocompatibility complex (MHC) class I region on chromosome 6.
Catalog No. 50-001-07842