Learn More
GFS Chemicals Inc 1,2-CYCLOHEXANEDIAMINETETRAACETIC ACID, REAGENT (ACS) | 100 G | GFS CHEMICALS, INC. Item #: 32301 | Bulk #: 688
SDP
Supplier:  GFS Chemicals Inc 32301
Encompass_Preferred
1,2-CYCLOHEXANEDIAMINETETRAACETIC ACID, REAGENT (ACS) | Chemical Formula : C!1!4H!2!2N!2O!8.H!2O | CAS # : 125572-95-4 | Specifications : Assay 97.5 - 100.5 %, Heavy metals (as Pb) 0.001 % , Insoluble in dil. NaOH (%) 0.01 % , Iron 0.005 % , Residue after ignition 0.2 %
Catalog No. NC9727166