Learn More
Hach Company CDTA Disodium Magnesium Salt Powder Pillows, pk/100
Supplier:  Hach Company 1408099
Encompass
CDTA Disodium Magnesium Salt. For interference removal in hardness titration tests.
Catalog No. 50-199-4923