Learn More
Hach Company CDTA Titrant (HexaVer™), 0.02 N, 4 L
Supplier: Hach Company 74056
CDTA Standard Solution, (HexaVer™), 0.020 N (N/50) (0.010 M) For hardness determination by buret titration. 4 liter bottle