Learn More
Hach Company CDTA Titrant (HexaVer™), 0.02 N, 1 L
Supplier:  Hach Company 74053
Encompass
CDTA Standard Solution, (HexaVer™), 0.020 N (N/50) (0.010 M) For hardness determination by buret titration. 1 liter bottle
Catalog No. 50-199-4440