Learn More
Nova Biomedical Corporation CDV REAGENT CARTRIDGE
Catalog No. NC0249896
Encompass
BioProfile Cell Density and Viability (CDV) are easy-to-use reagent cartridges analyze over 300 samples per cartridge and can be replaced in minutes.
Catalog No. NC0249896 Supplier Nova Biomedical Corporation Supplier No. 48796
BioProfile Cell Density and Viability (CDV) are easy-to-use reagent cartridges analyze over 300 samples per cartridge and can be replaced in minutes.