Learn More
Aviva Systems Biology Corp Cdx1 Antibody - C-terminal region (ARP38076_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38076_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cdx1. It was validated on Western Blot. . Alternate names: Cdx, Cdx-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-713