Learn More
Aviva Systems Biology Corp CDX1 Antibody - N-terminal region (ARP38075_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38075_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CDX1. Alternate names: MGC116915ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Zebrafish
Catalog No. 50-284-712