Learn More
Biogenex Laboratories CDX2
SDP
Supplier:  Biogenex Laboratories MU392AUC
Encompass
Anti-Human CDX-2; Clone: CDX2-88; Species Reactivity: Mouse; Isotype: IgG 1kappa; 1mL; Storage: 2 deg. to 8 deg.C
Catalog No. NC9471689