Learn More
enQuire Bioreagents CDX2 anti-Human Monoclonal [CDX2/1690], Size: 100ug
Supplier:  enQuire Bioreagents 1045MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: CDX2/1690. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HT29 cells or Colon Carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human CDX2 protein fragment (aa150-249). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 40kDa.Chromosome: 13q12.2. Gene Symbol: CDX2. Gene ID: 1045. Ensemble: ENSG00000165556. Uniprot: XP_011533178.
Catalog No. 50-188-4570