Learn More
Aviva Systems Biology Corp CDY1 Antibody - C-terminal region (ARP48645_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48645_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CDY1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CDY1. Alternate names: CDY, CDY1AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-59879