Learn More
Aviva Systems Biology Corp CDYL Antibody - N-terminal region (ARP49096_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49096_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CDYL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human CDYL. Alternate names: CDYL1, DKFZP586C1622, MGC131936ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60019