Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CE(20:0) (Cholesteryl arachidate) | 2573-03-7 | 99.9% | 681.17 | 250 MG
SDP

Catalog No. 5000438817
Encompass_Preferred

CE(20:0) (Cholesteryl arachidate) is an ester product intended for research use only. It is not for patient administration.

  • Solid appearance
  • White to off-white color
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C

Catalog No. 50-004-38817 Supplier Medchemexpress LLC Supplier No. 250MGHY113170
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CE(20:0) (Cholesteryl arachidate) is an ester product intended for research use only. It is not for patient administration.

  • Solid appearance
  • White to off-white color
  • Stable for 3 years as powder at -20°C
  • Stable for 6 months in solvent at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.