Learn More
TARGETMOL CHEMICALS INC CE-224535 5MG
SDP
Catalog No. 502581269
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CE-224535 (PF-04905428) is a selective antagonist of P2X7 receptor. CE-224535 can be used in disease-modifying antirheumatic studies. purity: 98%
Catalog No. 50-258-1269 Supplier TARGETMOL CHEMICALS INC Supplier No. T149205MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CE-224535 (PF-04905428) is a selective antagonist of P2X7 receptor. CE-224535 can be used in disease-modifying antirheumatic studies. purity: 98%