Learn More
Vileda Professional™ CE Duo™ CleanTech™ Sterile Mop with Bucket Liner
Catalog No. 19200841 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
Catalog No. Autoclavable
19-200-841 Not Autoclavable
1 options
Catalog No. 19-200-841 Supplier Vileda Professional™ Supplier No. 169581
Add to Cart
Add to Cart
Single-use, double-sided microfiber cleanroom foam mop for cleaning and disinfecting floors, walls and ceilings. Now available in a convenient 6 pack with included bucket liner.
  • Microfiber, overstitched cleanroom foam for cleaning/disinfecting floors, walls, and ceilings
  • Adhesive free lamination techniques are used to prevent additional contamination
  • Excellent microfiber cleaning results have been independently proven
  • Validated to 10-6 SAL per ISO 11137
  • Packaged 6 sterile mops per pack, double bagged with bucket liner
  • Reduce packaging waste
  • Reduce amount of time needed to pre-prepare the mops
Specifications
Product Type Double Sided Foam Mop
Sterility Sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 6 Pk, 8 Pk/Cs.
Includes Bucket Liner
For Use With (Application) Floor, Wall and Ceiling Cleaning and Disinfecting
Certifications/Compliance Certificate of Compliance
Material Microfiber