Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional CE isoXclean™ Isolator Tool

Catalog No. 17549303 Shop All Vileda Professional Products
Change view
Click to view available options
Sterility:
Non-sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17-549-303 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-549-303 Supplier Vileda Professional Supplier No. 182622
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lightweight ergonomic isolator cleaning tool with precision reach and microfiber for fast, consistent contamination removal in controlled environments.

  • Telescopic handle with an articulating, push-button activated, pivoting joint, allowing precise cleaning pad placement and movement in restricted spaces
  • Flexible mop handle and hinged frame are engineered for touchless-off mop pads without direct contact with operator
  • Maintains controlled, precise, contact with internal surfaces
  • Improves cleaning consistency, efficacy, and speed in highly regulated areas
  • Improved ergonomics
  • Time and cost efficiency- faster cleaning and pad changeover functionality reduces downtime between cleaning cycles
  • Application area: isolators, biosafety cabinets, RABS, and other hard to clean areas

Specifications

Product Type Isolator Cleaning Tool
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Reusable
Color-coded No
Material Polypropylene, Polyamide (Non-woven), Thermoplastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.