Learn More
Vileda Professional™ CE MicroIntensive Duo™
Catalog No. 19200828 Shop All Vileda Professional Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-200-828 Sterile
19-200-827 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-200-828 Supplier Vileda Professional™ Supplier No. 166815
Add to Cart
Add to Cart
Double-sided, single-use mop to speed-up the cleaning process, while also reducing the risk of cross-contamination.
  • Independent lab study, conducted by BMA Labs, resulting in bacteria reduction of 99.99%
  • 4-layer construction - featuring Polyester/Microfiber embossed cleaning side
  • Pre-laundered for single-use
  • Gamma and autoclave compatible
  • Extremely low particle shedding
  • Double-sided cleaning with color -stitched side indicator
  • Corner drainage openings for cleaner rinses
  • Finger loop for touchless on/off of cleaning sides
Specifications
Product Type Cleanroom Mop
Color White
Sterility Sterile
Autoclavable Autoclavable
Material Microfiber Polyester