Learn More
Vileda Professional CE MicroIntensive Duo™ Plus Mop
Catalog No. 17010401 Shop All Vileda Professional Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
17-010-401 Sterile
17-010-402 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-401 Supplier Vileda Professional Supplier No. 172300
Add to Cart
Add to Cart
Single-use mop for IPA rinsing and the removal of disinfectant residue on glass and stainless steel.
  • High performance dimpled polyester microfiber for excellent cleaning results
  • High surface coverage with double-sided design
  • Effective residue removal on glass and stainless steel
  • IPA compatible
  • Ideal for damp mopping and pre-preperation cleaning methods
  • Streak free cleaning
  • Extremely low particle shedding - Up to ISO 4 (GMP A/B)
  • Saves time, money, and chemical use
  • Gamma Irradiated
Specifications
Product Type Cleanroom Mop
Color Code White
Sterility Sterile
Autoclavable Autoclavable
Disposable Single-use
Color-coded No
Material Microfiber Polyester