Learn More
Vileda Professional CE MicroStrong™ Duo
Catalog No. 17103080 Shop All Vileda Professional Products
Change view
Click to view available options
Material:
Knitted Polyester Microfiber
Catalog No. Material
17-103-080 Knitted Polyester Microfiber
1 options
Catalog No. 17-103-080 Supplier Vileda Professional Supplier No. 182772
Add to Cart
Add to Cart
Double-sided, high-performance microfiber mop heads designed for controlled-environment and cleanroom cleaning
  • Designed for specialty cleaning of structured floors
  • High strength compact structure mop
  • Low particle shedding
  • Heat and chemical resistant
  • Suitable for gamma and autoclave sterilization
  • Suitable for ISO Class 4-9 / GMP A/B, C, D
Specifications
Product Type Mop Cover
Sterility Non-sterile
Autoclavable Autoclavable
Color-coded No
Material Knitted Polyester Microfiber