Learn More
Vileda Professional CE Pre-Prep Box Rack
Catalog No. 17103078 Shop All Vileda Professional Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
17-103-078 Stainless Steel
1 options
Catalog No. 17-103-078 Supplier Vileda Professional Supplier No. 179010
Add to Cart
Add to Cart
A reusable container used in controlled-environment cleaning workflows and designed to sit on trolleys
  • Designed to add compatibility of two pre-prep boxes to the Double Bucket Trolley
  • Easy addition to existing trolley
  • Specific to cleanroom cleaning
  • Fully autoclavable and chemical resistant
  • High durability and stability
Specifications
Product Type Bucket Attachment
Material Stainless Steel
Autoclavable Autoclavable
Disposable Reusable