Learn More
AB Sciex LLC CE-SDS Protein Analysis Kit, 200 tests, BioPhase 8800 system compatible, Capillary Electrophoresis-Sodium Dodecyl Sulfate assay, includes CE-SDS separation buffer, 10 kD internal standard
Catalog No. NC2157545
Encompass
The CE-SDS Protein Analysis Kit is designed for automated capillary electrophoresis-sodium dodecyl sulfate (CE-SDS) protein analysis on the BioPhase 8800 system. This kit enables the resolution of reduced and non-reduced proteins by size, allowing for comprehensive quantification of protein heterogeneity and impurities. It supports applications in biopharmaceutical analytical development and quality control.
  • Streamlines protein characterization workflows
  • High reproducibility and sensitive impurity detection
  • Provides trusted, quantitative data without enhancement
  • Supports compliant data for regulatory submissions
  • Offers superior resolution compared to traditional SDS-PAGE
  • Reduces hands-on time and operator variability
  • Configured for 200 tests per kit, optimizing high-throughput profiling
  • Includes necessary reagents and supplies for analysis
  • Research use only, not for diagnostic procedures
This item is not returnable. View return policy
Catalog No. NC2157545 Supplier AB Sciex LLC Supplier No. C30085
The CE-SDS Protein Analysis Kit is designed for automated capillary electrophoresis-sodium dodecyl sulfate (CE-SDS) protein analysis on the BioPhase 8800 system. This kit enables the resolution of reduced and non-reduced proteins by size, allowing for comprehensive quantification of protein heterogeneity and impurities. It supports applications in biopharmaceutical analytical development and quality control.
  • Streamlines protein characterization workflows
  • High reproducibility and sensitive impurity detection
  • Provides trusted, quantitative data without enhancement
  • Supports compliant data for regulatory submissions
  • Offers superior resolution compared to traditional SDS-PAGE
  • Reduces hands-on time and operator variability
  • Configured for 200 tests per kit, optimizing high-throughput profiling
  • Includes necessary reagents and supplies for analysis
  • Research use only, not for diagnostic procedures