Learn More
Bioventures Inc CE SIZING STANDARD 8FMOLES
Catalog No. 50272347
Encompass
MAPMARKER, 70-500BP, ROX LABELED (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400, 425, 450, 475, 490, 500bp) 8fmoles/band/ul Concentration
Catalog No. 50-272-347 Supplier Bioventures Inc Supplier No. MMFL 1000
MAPMARKER, 70-500BP, ROX LABELED (70, 80, 90, 100, 120, 140, 160, 180, 190, 200, 220, 240, 260, 280, 300, 320, 340, 360, 380, 400, 425, 450, 475, 490, 500bp) 8fmoles/band/ul Concentration