Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CE Tank Cleaner

Catalog No. 19200813 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-813 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-813 Supplier Vileda Professional™ Supplier No. 162234
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specialty above-the-floor tools are ideal for surfaces that are difficult to reach, clean and disinfect. Tools are easy to use and ergonomic allowing full range of motion during cleaning and disinfecting

  • Ideal for inner and outer tank walls
  • Manufactured of electro-polished stainless steel
  • Completely autoclavable
  • Frame is pre-bent to 9 in. radius but is easily adjusted, and can be flat to use on isolators
  • 4 degree of freedom to ensure good contact with the surface being cleaned
  • Easy to use with handle for tight spaces and can be used with telescopic handle for hard to reach areas

Specifications

Product Type Tank Cleaning Tool
Color Silver
Sterility Non-sterile
Autoclavable Autoclavable
Size 9 in.
Quantity 1
For Use With (Application) Inner and Outer Tanks Walls
Material Electropolished Stainless Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.