Learn More
Vileda Professional CE TraXero™ Base
Catalog No. 17549310 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
17-549-310 Gray
1 options
Catalog No. 17-549-310 Supplier Vileda Professional Supplier No. 182778
Add to Cart
Add to Cart
Main track unit of the TraXero™ system, enabling fast, consistent wheel and castor decontamination to prevent contamination in cleanroom environments
  • Designed for easy alignment and guidance of wheels
  • Modular design allows for add-on bases and gates to extend cleaning length
Specifications
Product Type Base
Color Gray