Learn More
Vileda Professional CE TraXero™ Replacement Gates
Catalog No. 17549313 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
17-549-313 Gray
1 options
Catalog No. 17-549-313 Supplier Vileda Professional Supplier No. 182854
Add to Cart
Add to Cart
Spare components that maintain the performance and reliability of the TraXero™ wheel cleaning system, ensures proper wheel guidance and consistent decontamination
  • CE TraXero™ Replacement Gates (Vileda Professional) are spare/consumable components used within the TraXero™ Wheel Cleaning system for controlled environments.
  • Honeycomb backer designed to allow faster dosing and dispersion of chemical cleaning agents
  • Gates provide rigidity needed for mechanical removal of particles
Specifications
Product Type Replacement Gates
Color Gray