Learn More
Vileda Professional CE TraXero™ Wheel Cleaner Kit
Catalog No. 17549314 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
Gray
Catalog No. Color
17-549-314 Gray
1 options
Catalog No. 17-549-314 Supplier Vileda Professional Supplier No. 182860
Add to Cart
Add to Cart
TraXero Wheel Cleaner Kit ensures fast, consistent wheel decontamination before cleanroom entry, reducing contamination risk and supporting compliance.
  • Two-track castor cleaning system is an automated solution for decontaminating trolley wheels and castors.
  • Trolly are guided through the track and gate assembly by the user.
  • Castors are cleaned by a microfiber gated system validated to ISO 4 standards.
Specifications
Product Type Wheel Cleaner Kit
Color Gray