Learn More
Vileda Professional CE TraXero™ Wheel Cleaner Pads - Gamma Irradiated
Catalog No. 17549312 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-549-312 White
1 options
Catalog No. 17-549-312 Supplier Vileda Professional Supplier No. 182814
Add to Cart
Add to Cart
Sterile gamma-irradiated microfiber pads for TraXero™, delivering consistent 4 log wheel cleaning and reliable contamination control
  • Disposable microfiber cleaning refill
  • Cleanroom packaged and poly-bagged
  • Symmetrical design so it can be placed into the gate on either side of TraXero™ tracks
  • Dark blue tabs on backing for ergonomic install and removal from gates
  • Suitable for ISO class 4-9 / GMP A/B, C
  • Chemical resistance against most disinfectant categories
  • Lot number, expiration date, and gamma irradiation indicator dot on each package
  • Certificate of Irradiation available with each shipment
Specifications
Product Type Wheel Cleaner Pads
Color White