Learn More
Vileda Professional CE TraXero™ Wheel Cleaner Pads
Catalog No. 17549311 Shop All Vileda Professional Products
Change view
Click to view available options
Color:
White
Catalog No. Color
17-549-311 White
1 options
Catalog No. 17-549-311 Supplier Vileda Professional Supplier No. 182813
Add to Cart
Add to Cart
Evolon™ microfiber pads deliver fast, consistent 4 log wheel cleaning, ensuring reliable particle removal and repeatable contamination control.
  • Disposable microfiber cleaning refill
  • Cleanroom packaged and poly-bagged
  • Symmetrical design so it can be placed into the gate on either side of TraXero™ tracks
Specifications
Product Type Wheel Cleaner Pads
Color White