Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CE UltraSpeed Pro™ Frame

Catalog No. 19200771 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-771 Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-771 Supplier Vileda Professional™ Supplier No. 157777
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The most effective and easy to use bucket and wringer system for use within Controlled Environments!

Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.

  • Lightweight, durable and autoclavable frame
  • 40cm/16in breakaway frame can be used with all UltraSpeed Pads
  • Frame includes a modern swivel joint with integrated ball catch making it easy to clean vertical surfaces
  • Easy and fast rinsing of mops without removal of frame
  • Stainless steel springs

Specifications

Color Blue
Length (English) 16 in.
Autoclavable Autoclavable
Quantity 10/Cs.
For Use With (Equipment) 119966, 157775, 157779
For Use With (Application) ISO 4 to 9/GMP A/B, C, D on Floors, Walls and Ceilings
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.