Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vileda Professional™ CE UltraSpeed Pro™ MicroIntensive™ L Mop

Catalog No. 19200772 Shop All Vileda Professional Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
19-200-772 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-200-772 Supplier Vileda Professional™ Supplier No. 157779
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The most effective and easy to use bucket and wringer system for use within Controlled Environments!

Demands on effectiveness, cost, ease of handling and staff training is constantly increasing. Whatever the cleaning requirements, the CE UltraSpeed Pro™ Double Bucket System will dramatically increase speed and cleaning performance while decreasing total costs and providing versatility and ergonomics.

  • High performance microfiber blend ideal for cleaning floors and walls
  • Independently proven bacteria reduction of 99.997%
  • Highly absorbent with even water release and easy rinsing
  • Very low particle shedding construction
  • High chemical resistance against most disinfectants
  • Pre-laundered and single bagged
  • Disposable

Specifications

Product Type Single Use Mop
Color White
Length (English) 16 in.
Sterility Non-sterile
Autoclavable Not Autoclavable
Disposable Single-use
Quantity 50/Cs.
For Use With (Equipment) 157777, 157775 and 119966
For Use With (Application) ISO 4 to 9/GMP A/B, C, D on Floors, Walls and Ceilings
Material Microfiber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.