Learn More
Medchemexpress LLC HY-108539 5mg Medchemexpress, CE3F4 CAS:143703-25-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1085395MG
Encompass_Preferred
Medchemexpress, HY-108539 5mg CE3F4 CAS:143703-25-7 CE3F4 is a selective antagonist of exchange protein directly activated by cAMP (Epac1), with IC50s of 10.7 μM and 66 μM for Epac1 and Epac2(B), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9635