Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CE3F4 analog 1 | 2141967-43-1 | 99.7% | C11H9Br3FNO | 10MG
SDP

Catalog No. 5000258276
Encompass_Preferred

CE3F4 analog 1 is a small-molecule analogue of CE3F4, a tribromo- and fluoro-substituted quinolinecarboxaldehyde used in research on Ras and GPCR/G-protein pathways. It is supplied as a solid and as DMSO solutions for biochemical and cellular studies, with stability and documentation provided for research use.

  • High purity: 99.7% (manufacturer specification).
  • Chemical formula C11H9Br3FNO; molecular weight 429.91 g/mol.
  • Appearance: solid, white to off-white.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available as solid masses and 10 mM solution in DMSO for assay convenience.
  • SMILES: O=CN1C(C)CCC2=C1C(Br)=C(Br)C(F)=C2Br.

Catalog No. 50-002-58276 Supplier Medchemexpress LLC Supplier No. HY13387510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CE3F4 analog 1 is a small-molecule analogue of CE3F4, a tribromo- and fluoro-substituted quinolinecarboxaldehyde used in research on Ras and GPCR/G-protein pathways. It is supplied as a solid and as DMSO solutions for biochemical and cellular studies, with stability and documentation provided for research use.

  • High purity: 99.7% (manufacturer specification).
  • Chemical formula C11H9Br3FNO; molecular weight 429.91 g/mol.
  • Appearance: solid, white to off-white.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Available as solid masses and 10 mM solution in DMSO for assay convenience.
  • SMILES: O=CN1C(C)CCC2=C1C(Br)=C(Br)C(F)=C2Br.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.