Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ce3f4 analog 1 | 2141967-43-1 | MFCD32857158 | 99.7% | 429.91 | C11H9Br3FNO | 5 MG
SDP

Catalog No. 5000262490
Encompass_Preferred

CE3F4 analog 1 is a small-molecule research compound used as an analogue of CE3F4 for biochemical and cell-based studies targeting Epac/Ras-related signaling. It is supplied as a high-purity solid and is also available as a DMSO solution for in-vitro experiments. Store under recommended cold conditions to maintain stability.

  • High purity (99.67%) suitable for analytical and biological assays.
  • Molecular weight 429.91 for accurate stoichiometry calculations.
  • CAS number 2141967-43-1 for unambiguous identification.
  • Available as small solid quantities and as a 10 mM solution in DMSO for convenience.
  • Recommended storage: powder at -20°C; solution at -80°C for long-term stability.
  • Intended for laboratory research use only; not for human or clinical use.

Catalog No. 50-002-62490 Supplier Medchemexpress LLC Supplier No. HY1338755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CE3F4 analog 1 is a small-molecule research compound used as an analogue of CE3F4 for biochemical and cell-based studies targeting Epac/Ras-related signaling. It is supplied as a high-purity solid and is also available as a DMSO solution for in-vitro experiments. Store under recommended cold conditions to maintain stability.

  • High purity (99.67%) suitable for analytical and biological assays.
  • Molecular weight 429.91 for accurate stoichiometry calculations.
  • CAS number 2141967-43-1 for unambiguous identification.
  • Available as small solid quantities and as a 10 mM solution in DMSO for convenience.
  • Recommended storage: powder at -20°C; solution at -80°C for long-term stability.
  • Intended for laboratory research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.