Learn More
CELL MARQUE CORPORATION CEA (CEA31) Mouse Monoclonal Antibody, 0.1 mL Concentrate
SDP
Supplier:  CELL MARQUE CORPORATION 236M94
Encompass
Anti-CEA is an antibody against carcinoembryonic antigen (CEA), a protein thought to promote tumor development through its role as a cell adhesion molecule. Anti-CEA positivity is seen in adenocarcinomas of many origins, especially colon and lung adenocarcinomas, but rarely seen in mesothelial cells and mesotheliomas
Catalog No. NC2746623