Learn More
MOLECULAR DEPOT LLC CEA mAb - 1 mg
Catalog No. 5000013504
Encompass_Preferred
CEA Monoclonal Antibody (1 mg) is an affinity-purified mouse monoclonal antibody specific to human Carcinoembryonic Antigen (CEA) a glycoprotein produced during the fetal stage of human development Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only
Catalog No. 50-000-13504 Supplier MOLECULAR DEPOT LLC Supplier No. A2010012
CEA Monoclonal Antibody (1 mg) is an affinity-purified mouse monoclonal antibody specific to human Carcinoembryonic Antigen (CEA) a glycoprotein produced during the fetal stage of human development Tested in several immunoassay platforms including ELISA CLIA Immunoturbidimetry and more tested matching clones are available Readily available in bulk amounts and at bulk prices Biotinylation and other labeling services are also available For in vitro research use only