Learn More
Genscript Corporation CEACAM-5/CD66eBiotin His
Catalog No. 5000057149
Encompass
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) was identified as a metastatic driver CEACAM5 overproduction enriched for an epithelial gene expression pattern and facilitated tumor outgrowth at metastatic sites Tissues from patients with metastatic breast cancer confirmed elevated levels of CEACAM5 in lung metastases relative to breast tumors and an inverse correlation between CEACAM5 and the mesenchymal marker vimentin was demonstrated
Catalog No. 50-000-57149 Supplier Genscript Corporation Supplier No. Z03834100
Carcinoembryonic antigen-related cell adhesion molecule 5 (CEACAM5) was identified as a metastatic driver CEACAM5 overproduction enriched for an epithelial gene expression pattern and facilitated tumor outgrowth at metastatic sites Tissues from patients with metastatic breast cancer confirmed elevated levels of CEACAM5 in lung metastases relative to breast tumors and an inverse correlation between CEACAM5 and the mesenchymal marker vimentin was demonstrated