Learn More
Genscript Corporation CEACAM-6/CD66cBiotin His A
Catalog No. 5000056964
Encompass
Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6) belongs to the human carcino-embryonic antigen (CEA) family Numerous lines of studies have indicated that altered expression of CEACAM6 may have a role in carcinogenesis and development
Catalog No. 50-000-56964 Supplier Genscript Corporation Supplier No. Z03837500
Carcinoembryonic antigen-related cell adhesion molecule 6 (CEACAM6) belongs to the human carcino-embryonic antigen (CEA) family Numerous lines of studies have indicated that altered expression of CEACAM6 may have a role in carcinogenesis and development