Learn More
ACROBiosystems ACROBiosystems Biotinylated Human CEACAM-6 / CD66c Protein, His,Avitag™
Supplier:  ACROBiosystems CE6H82E7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human CEACAM-6, His,Avitag (CE6-H82E7) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession # P40199-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.9 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CEACAM-8, His Tag (Cat. No. CE8-H5224) at 5 μg/mL (100 μL/well) can bind Biotinylated Human CEACAM-6, His,Avitag (Cat. No. CE6-H82E7) with a linear range of 0.078-1.25 μg/mL (QC tested).
Catalog No. 50-195-7091