Learn More
Genscript Corporation CEACAM-8/CD66bBiotin His
Catalog No. 5000059094
Encompass
Eosinophils and their products are likely important in the pathophysiology of allergic diseases such as bronchial asthma and in host immunity to parasitic organisms CD66b (CEACAM8 CGM6 NCA-95) is a single chain GPI-anchored highly glycosylated protein belonging to the carcinoembryonic Ag supergene family CD66b is an activation marker for human granulocytes
Catalog No. 50-000-59094 Supplier Genscript Corporation Supplier No. Z04432100
Eosinophils and their products are likely important in the pathophysiology of allergic diseases such as bronchial asthma and in host immunity to parasitic organisms CD66b (CEACAM8 CGM6 NCA-95) is a single chain GPI-anchored highly glycosylated protein belonging to the carcinoembryonic Ag supergene family CD66b is an activation marker for human granulocytes