Learn More
enQuire Bioreagents Carcinoembryonic Antigen / CD66 Anti-Human Monoclonal [Clone C66/195]; Size: 100ug
Supplier:  enQuire Bioreagents 1048MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: C66/195. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: MCF7 or 293T cells. Colon carcinoma. Cellular Staining: Cytoplasmic, luminal surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant full-length human CEA protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 80-200kDa.Chromosome: 19q13.1-19q13.2. Gene Symbol: CEACAM1 & CEACAM5. Gene ID: 634. Ensemble: ENSG00000079385. Uniprot: NP_011525508.
Catalog No. 50-188-4573