Learn More
Enzo Life Sciences CEACAM1 (human) monoclonal antibody (GM8G5) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805076C100
Encompass_Preferred
Clone: GM8G5. Isotype: IgG1. Immunogen: Vector containing the cDNA of human CEACAM1 A2 domain. Species reactivity: Human. Applications: Flow Cytometry, IHC (FS). Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2. Contains no preservatives.
Catalog No. 50-201-0051