Learn More
Medchemexpress LLC CEACAM1 Human N-Hi 10ug
SDP
Catalog No. 5000196266
Encompass_Preferred
Recombinant human CEACAM1 protein with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder. The protein is suitable for biochemical and cell signaling studies involving insulin receptor-mediated pathways, hepatic lipogenesis, and modulation of cell proliferation. Supplied as 10 μg with >95% purity and low endotoxin, it includes recommended storage and reconstitution instructions.
  • Expressed in E. coli for high-yield production.
  • N-terminal His tag enabling straightforward purification and detection.
  • Lyophilized format for convenient storage and transport.
  • High purity for reliable biochemical assays.
  • Low endotoxin to support cell-based experiments.
  • Stable when aliquoted and frozen for long-term storage.
Catalog No. 50-001-96266 Supplier Medchemexpress LLC Supplier No. HYP72129A10UG
Recombinant human CEACAM1 protein with an N-terminal His tag, expressed in E. coli and supplied as a lyophilized powder. The protein is suitable for biochemical and cell signaling studies involving insulin receptor-mediated pathways, hepatic lipogenesis, and modulation of cell proliferation. Supplied as 10 μg with >95% purity and low endotoxin, it includes recommended storage and reconstitution instructions.
  • Expressed in E. coli for high-yield production.
  • N-terminal His tag enabling straightforward purification and detection.
  • Lyophilized format for convenient storage and transport.
  • High purity for reliable biochemical assays.
  • Low endotoxin to support cell-based experiments.
  • Stable when aliquoted and frozen for long-term storage.