Learn More
Enzo Life Sciences CEACAM19 (human) monoclonal antibody (HY-8H10) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805089C100
Encompass_Preferred
Clone: HY-8H10. Isotype: IgG1. Immunogen: Vector containing the cDNA of human CEACAM19. Species reactivity: Human. Applications: ELISA, Flow Cytometry. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS, pH 7.2. Contains no preservatives.
Catalog No. 50-201-0056