Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human CEACAM-1 / CD66a Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;CE1,CEACAM1,CD66a,BGP,BGP1,BGP-1,BGPI
Supplier:  ACROBiosystems CE1H82E525UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human CEACAM-1 / CD66a Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CEACAM-1, His Tag (CE1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gly 428 (Accession # P13688).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 45.9 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8224